missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one, 98%, Thermo Scientific™

Product Code. 15419005
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419005 250 mg 250mg
15409005 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419005 Supplier Thermo Scientific Alfa Aesar Supplier No. H35321.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421263-65-1
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.19
MDL Number MFCD22988925
InChI Key AMETXLSIVXCORJ-UHFFFAOYSA-N
Synonym 6,7-dihydrocyclopenta 4,5 pyrrolo 3,2-b pyridin-8 5h-one, 7,12-diazatricyclo 6.4.0.0 2 ,? dodeca-1 12 ,2 6 ,8,10-tetraen-3-one
PubChem CID 73995189
IUPAC Name 6,7-dihydro-5H-cyclopenta[2,3]pyrrolo[2,4-b]pyridin-8-one
SMILES O=C1CCC2=C1C1=NC=CC=C1N2

Specifications

Color Green
Quantity 250 mg
Formula Weight 172.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 6,7-Dihydrocyclopenta[4,5]pyrrolo[3,2-b]pyridin-8(5H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.