missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,7-Dihydro Rosuvastatin Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 6,7-Dihydro Rosuvastatin sodium salt |
| CAS | 1347362-67-7 |
| Molecular Formula | C22 H29 F N3 O6 S . Na |
| Quantity | 100 mg |
| Synonym | 5-Pyrimidineheptanoic acid, 4-(4-fluorophenyl)-β,Δ-dihydroxy-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-, sodium salt (1:1), (βR,ΔR)-, (βR,ΔR)-4-(4-Fluorophenyl)-β,Δ-dihydroxy-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidineheptanoic acid sodium salt (1:1), (3R,5R)-7-[4-(4-Fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-3,5-dihydroxyheptanoic acid sodium salt, 6,7-Dihydro Rosuvastatin sodium salt, 6,7-Dihydrorosuvastatin sodium |
| InChI Formula | InChI=1S/C22H30FN3O6S.Na/c1-13(2)20-18(10-9-16(27)11-17(28)12-19(29)30)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)33(4,31)32;/h5-8,13,16-17,27-28H,9-12H2,1-4H3,(H,29,30);/q;+1/p-1/t16-,17-;/m1./s1 |
| SMILES | [Na+].CC(C)c1nc(nc(c1CC[C@@H](O)C[C@@H](O)CC(=O)[O-])c2ccc(F)cc2)N(C)S(=O)(=O)C |
| IUPAC Name | sodium;(3R,5R)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyheptanoate |
| Molecular Weight (g/mol) | 505.54 |
| Formula Weight | 505.1659 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?