missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydro-4(5H)-benzofuranone, 98%

Product Code. 10365271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10365271 5 g Glass bottle 5g
10738041 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10365271 Supplier Thermo Scientific Acros Supplier No. 330080050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16806-93-2 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16806-93-2
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00215511
InChI Key DXWQOYPYNPSVRL-UHFFFAOYSA-N
Synonym 6,7-dihydro-4 5h-benzofuranone, 6,7-dihydrobenzofuran-4 5h-one, 4,5,6,7-tetrahydro-1-benzofuran-4-one, 6,7-dihydro-1-benzofuran-4 5h-one, 4 5h-benzofuranone, 6,7-dihydro, pubchem14044, 6,7-dihydro-4 5h benzo-furanone, 6,7-dihydro-5h-benzofuran-4-one, 6?7-dihydro-4 5h-benzofuranone, 6,7-dihydro-4-5h-benzofuranone
PubChem CID 2733541
IUPAC Name 6,7-dihydro-5H-1-benzofuran-4-one
SMILES C1CC2=C(C=CO2)C(=O)C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 30.0°C to 34.0°C
Color Yellow to Brown
Boiling Point 115.0°C to 118.0°C (16.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5300 to 1.5320
Quantity 5 g
Beilstein 17, V,9, 585
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methylene chloride,ether
Formula Weight 136.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 6, 7-Dihydro-4(5H)-benzofuranone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.