missing translation for 'onlineSavingsMsg'
Learn More

6,7,8,9-Tetrahydro-5h-carbazole-3-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10357223
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10357223 1 g 1g
10505554 250 mg 250mg
10461124 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10357223 Supplier Thermo Scientific Maybridge Supplier No. MO07745DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 36729-27-8
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD00458819
InChI Key OWQQDAGRTDUORV-UHFFFAOYSA-N
Synonym 2,3,4,9-tetrahydro-1h-carbazole-6-carboxylic acid, 1,2,3,4-tetrahydrocarbazole-6-carboxylic acid, 1h-carbazole-6-carboxylic acid, 2,3,4,9-tetrahydro, 5,6,7,8,9-pentahydro-4ah-carbazole-3-carboxylic acid, enamine_005086, 2,3,4,9-tetrahydro-1h-carbazole-6-carboxylicacid, 1,2,3,4-tetrahyrocarbazole-6-carboxylic acid
PubChem CID 729801
IUPAC Name 6,7,8,9-tetrahydro-5H-carbazole-3-carboxylic acid
SMILES C1CCC2=C(C1)C3=C(N2)C=CC(=C3)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.25
Percent Purity 97%
Chemical Name or Material 6,7,8,9-Tetrahydro-5h-carbazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.