missing translation for 'onlineSavingsMsg'
Learn More

6,6’-Methylenebis[(3RS)-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-one(Ondansetron Impurity B), TRC

Product Code. 30478649
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478649 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478649 Supplier Toronto Research Chemicals Supplier No. TRCM3039505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6,6'-Methylenebis[(3RS)-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-one]
CAS 1076198-52-1
Molecular Formula C37 H38 N6 O2
Quantity 5 mg
Synonym 6,6'-Methylenebis[(3RS)-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-one], Ondansetron Hydrochloride Dihydrate Imp. B (EP)
InChI Formula InChI=1S/C37H38N6O2/c1-22-38-13-15-42(22)20-26-7-11-32-34(36(26)44)28-18-24(5-9-30(28)40(32)3)17-25-6-10-31-29(19-25)35-33(41(31)4)12-8-27(37(35)45)21-43-16-14-39-23(43)2/h5-6,9-10,13-16,18-19,26-27H,7-8,11-12,17,20-21H2,1-4H3
SMILES Cc1nccn1CC2CCc3c(C2=O)c4cc(Cc5ccc6c(c5)c7C(=O)C(Cn8ccnc8C)CCc7n6C)ccc4n3C
IUPAC Name 9-methyl-3-[(2-methylimidazol-1-yl)methyl]-6-[[9-methyl-6-[(2-methylimidazol-1-yl)methyl]-5-oxo-7,8-dihydro-6H-carbazol-3-yl]methyl]-2,3-dihydro-1H-carbazol-4-one
Molecular Weight (g/mol) 598.74
Formula Weight 598.3056
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.