missing translation for 'onlineSavingsMsg'
Learn More

6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione, 98%, Thermo Scientific Chemicals

Product Code. 15460107
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67911-21-1
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD09751198
InChI Key QKAHKEDLPBJLFD-UHFFFAOYSA-N
PubChem CID 10374536
IUPAC Name 6,6-dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
SMILES CC1(C2C1C(=O)OC2=O)C

Specifications

Melting Point 53°C to 57°C
Color White
Boiling Point 137°C (14 mmHg)
Quantity 25 g
Beilstein 117391
Sensitivity Moisture sensitive
Synonym 6,6-dimethyl-3-oxabicyclo 3.1.0 hexane-2,4-dione, caronic anhydride, 3-oxabicyclo 3.1.0 hexane-2,4-dione, 6,6-dimethyl, 3,3-dimethyl-1,2-cyclopropanedicarboxylic anhydride, ksc626s3r, 6,6-dimethyl-3-oxabicyclo 3.1.0 hex-2,4-dione, 3,3-dimethyl-1,2-cyclopropane dicarboxylic anhydride, 3-oxabicyclo 3.1.0 hexane-2,4-dione,6,6-dimethyl, 3,3-dimethyl-1,2-cyclopropanedicarboxylic acid anhydride, 2r,4r-methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride form
Formula Weight 140.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'