missing translation for 'onlineSavingsMsg'
Learn More

6,6'-Dimethoxy-2,2'-binaphthalene, TRC

Product Code. 30379309
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379309 25 mg
30381730 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379309 Supplier Toronto Research Chemicals Supplier No. TRCD46136525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29619-45-2
Molecular Formula C22 H18 O2
Molecular Weight (g/mol) 314.38
Synonym 6,6'-Dimethoxy-2,2'-binaphthalenyl, Nabumetone Imp. F (EP)
IUPAC Name 2-methoxy-6-(6-methoxynaphthalen-2-yl)naphthalene
SMILES COc1ccc2cc(ccc2c1)c3ccc4cc(OC)ccc4c3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H18O2/c1-23-21-9-7-17-11-15(3-5-19(17)13-21)16-4-6-20-14-22(24-2)10-8-18(20)12-16/h3-14H,1-2H3
Formula Weight 314.1307
Chemical Name or Material 6,6'-Dimethoxy-2,2'-binaphthalenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.