missing translation for 'onlineSavingsMsg'
Learn More

6,6'-Dihydroxypregabalin Lactam, TRC

Product Code. 30382785
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382785 1 mg 1mg
30374666 10 mg 10mg
30382546 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382785 Supplier Toronto Research Chemicals Supplier No. TRCD4546451MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6,6'-Dihydroxypregabalin Lactam
Analyte or Component Names 6,6'-Dihydroxypregabalin Lactam
Shipping Condition Room Temperature
Molecular Formula C8H15NO3
Synonym 4-(3-Hydroxy-2-(hydroxymethyl)propyl)pyrrolidin-2-one
InChI Formula InChI=1S/C14H11ClN2O/c1-18-9-3-5-12-11(7-9)14(16)10-4-2-8(15)6-13(10)17-12/h2-7H,1H3,(H2,16,17)
Solution Type Neat
SMILES OCC(CO)CC1CNC(=O)C1
Industry Type Pharmaceutical
Molecular Weight (g/mol) 173.21
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.