missing translation for 'onlineSavingsMsg'
Learn More

6,6″-Dichloro-3,2':4',3″-terpyridine, 95%, Thermo Scientific™

Product Code. 11486500
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
11486500 100 mg 100mg
11496500 500 mg 500mg
2 options
This item is not returnable. View return policy

Product Code. 11486500

Brand: Thermo Scientific Alfa Aesar H27593.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942206-24-8
Molecular Formula C15H9Cl2N3
Molecular Weight (g/mol) 302.158
MDL Number MFCD09037443
InChI Key CWMMDFDDGLQUBF-UHFFFAOYSA-N
Synonym 6,6'-dichloro-3,2':4',3'-terpyridine, 2,4-bis 6-chloropyridin-3-yl pyridine, 6,6-dichloro-3,2':4',3-terpyridine, 6,6-dichloro-3,2':4',3'-terpyridine, 1∼6∼,3∼6∼-dichloro-1∼3∼,2∼2∼:2∼4∼,3∼3∼-terpyridine, 6'-chloro-4-6-chloropyridin-3-yl-2,3'-bipyridine
PubChem CID 42553048
IUPAC Name 2,4-bis(6-chloropyridin-3-yl)pyridine
SMILES C1=CC(=NC=C1C2=CC(=NC=C2)C3=CN=C(C=C3)Cl)Cl

Specifications

Quantity 100 mg
Formula Weight 302.16
Percent Purity 95%
Chemical Name or Material 6,6''-Dichloro-3,2':4',3''-terpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.