missing translation for 'onlineSavingsMsg'
Learn More

6-[6-(Carboxyamino)hexanamido]-hexanoic Acid N-Benzyl Methyl Ester, TRC

Product Code. 30011720
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011720 500 mg 500mg
30005662 1 g 1g
30010291 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011720 Supplier Toronto Research Chemicals Supplier No. TRCC177788500MG
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25580-88-5
Molecular Formula C21 H32 N2 O5
Molecular Weight (g/mol) 392.489
Synonym 6-[[1-Oxo-6-[[(phenylmethoxy)carbonyl]amino]hexyl]amino]-hexanoic Acid Methyl Ester
IUPAC Name methyl 6-[6-(phenylmethoxycarbonylamino)hexanoylamino]hexanoate
SMILES COC(=O)CCCCCNC(=O)CCCCCNC(=O)OCc1ccccc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H32N2O5/c1-27-20(25)14-8-4-9-15-22-19(24)13-7-3-10-16-23-21(26)28-17-18-11-5-2-6-12-18/h2,5-6,11-12H,3-4,7-10,13-17H2,1H3,(H,22,24)(H,23,26)
Formula Weight 392.231
Chemical Name or Material 6-[6-(Carboxyamino)hexanamido]-hexanoic Acid N-Benzyl Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.