missing translation for 'onlineSavingsMsg'
Learn More

6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester, Thermo Scientific™

Product Code. 11414620
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414620 250 mg 250mg
11424620 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414620 Supplier Thermo Scientific Alfa Aesar Supplier No. H50143.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485799-04-0
Molecular Formula C15H23BN2O3
Molecular Weight (g/mol) 290.17
MDL Number MFCD04039874
InChI Key ZGDLVKWIZHHWIR-UHFFFAOYSA-N
Synonym 4-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 6-morpholin-4-yl pyridine-3-boronic acid pinacol ester, 6-morpholinopyridin-3-ylboronic acid pinacol ester, 2-morpholinopyridine-5-boronic acid, pinacol ester, 4-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-morpholine, 4-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl morpholine, 2-4-morpholino pyridine-5-boronic acid pinacol ester, 4-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 6-morpholinopyridine-3-boronic acid pinacol ester
PubChem CID 2795363
IUPAC Name 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(N=C1)N1CCOCC1

Specifications

Quantity 250 mg
Formula Weight 290.17
Chemical Name or Material 6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.