missing translation for 'onlineSavingsMsg'
Learn More

6-(4-(5-Chloropentanamido)phenyl)-1-(4-methoxyphenyl)-7-oxo-6,7-dihydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic Acid Ethyl Ester, TRC

Product Code. 30361449
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361449 10 mg 10mg
30355145 25 mg -
30368032 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361449 Supplier Toronto Research Chemicals Supplier No. TRCC37121010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C27H27ClN4O5
Molecular Weight (g/mol) 522.98
IUPAC Name ethyl 6-(4-(5-chloropentanamido)phenyl)-1-(4-methoxyphenyl)-7-oxo-6,7-dihydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate
SMILES O=C(C1=NN(C2=CC=C(OC)C=C2)C3=C1C=CN(C4=CC=C(NC(CCCCCl)=O)C=C4)C3=O)OCC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H27ClN4O5/c1-3-37-27(35)24-22-15-17-31(19-9-7-18(8-10-19)29-23(33)6-4-5-16-28)26(34)25(22)32(30-24)20-11-13-21(36-2)14-12-20/h7-15,17H,3-6,16H2,1-2H3,(H,29,33)
Formula Weight 522.17
Chemical Name or Material 6-(4-(5-Chloropentanamido)phenyl)-1-(4-methoxyphenyl)-7-oxo-6,7-dihydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.