missing translation for 'onlineSavingsMsg'
Learn More

6,2'-Dihydroxyflavone, MedChemExpress

Product Code. 16484947
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16484947 Supplier Medchem Express Supplier No. HYN6628

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 6,2'-Dihydroxyflavone is a novel antagonist of GABAA receptor.

6,2'-Dihydroxyflavone is a novel antagonist of GABAA receptor.

Specifications

Chemical Name or Material 6,2'-Dihydroxyflavone
CAS 92439-20-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H10O4
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (393.33 mM)
SMILES O=C1C=C(C2=CC=CC=C2O)OC3=CC=C(O)C=C13
Molecular Weight (g/mol) 254.24
Formula Weight 254.24
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.