missing translation for 'onlineSavingsMsg'
Learn More

6-[2-Biotinylamidoethyl]-dithiopropionamido]-4,8-diaza-5,7-diketoundecanoic Acid Bis-N-sulfosuccinimidyl Ester Disodium Salt, TRC

Product Code. 30340309
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340309 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340309 Supplier Toronto Research Chemicals Supplier No. TRCB3927005MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-[2-Biotinylamidoethyl]-dithiopropionamido]-4,8-diaza-5,7-d
CAS 1044220-56-5
Molecular Formula C32 H42 N8 O19 S5 . 2 Na
Quantity 5 mg
Synonym 6-[2-Biotinylamidoethyl]-Dithiopropionamido]-4,8-Diaza-5,7-Diketoundecanoic Acid, Bis-N-Sulfosuccinimidyl Ester Disodium Salt
InChI Formula InChI=1S/C32H44N8O19S5.2Na/c41-20(4-2-1-3-17-26-16(15-60-17)36-32(51)38-26)33-10-12-62-61-11-7-21(42)37-27(28(47)34-8-5-24(45)58-39-22(43)13-18(30(39)49)63(52,53)54)29(48)35-9-6-25(46)59-40-23(44)14-19(31(40)50)64(55,56)57;;/h16-19,26-27H,1-15H2,(H,33,41)(H,34,47)(H,35,48)(H,37,42)(H2,36,38,51)(H,52,53,54)(H,55,56,57);;/q;2*+1/p-2/t16-,17-,18?,19?,26-,27?;;/m0../s1
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1CC(=O)N(OC(=O)CCNC(=O)C(NC(=O)CCSSCCNC(=O)CCCC[C@@H]2SC[C@@H]3NC(=O)N[C@H]23)C(=O)NCCC(=O)ON4C(=O)CC(C4=O)S(=O)(=O)[O-])C1=O
IUPAC Name disodium;1-[3-[[2-[3-[2-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]ethyldisulfanyl]propanoylamino]-3-[[3-(2,5-dioxo-3-sulfonatopyrrolidin-1-yl)oxy-3-oxopropyl]amino]-3-oxopropanoyl]amino]propanoyloxy]-2,5-dioxopyrrolidine-3-sulfonate
Molecular Weight (g/mol) 1049.02
Formula Weight 1048.0965
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.