missing translation for 'onlineSavingsMsg'
Learn More

6-[[2-(3,4-Dimethoxyphenyl)ethyl]amino]-6-oxo-hexanoic Acid Methyl Ester, TRC

Product Code. 30388655
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388655 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388655 Supplier Toronto Research Chemicals Supplier No. TRCD454425500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-((2-(3,4-Dimethoxyphenyl)ethyl)amino)-6-oxo-hexanoic Acid Methyl Ester
CAS 1435490-39-3
Molecular Formula C17H25NO5
Quantity 500 mg
InChI Formula InChI=1S/C17H25NO5/c1-21-14-9-8-13(12-15(14)22-2)10-11-18-16(19)6-4-5-7-17(20)23-3/h8-9,12H,4-7,10-11H2,1-3H3,(H,18,19)
SMILES COC(=O)CCCCC(=O)NCCc1ccc(OC)c(OC)c1
IUPAC Name methyl 6-[2-(3,4-dimethoxyphenyl)ethylamino]-6-oxohexanoate
Molecular Weight (g/mol) 323.38
Formula Weight 323.1733
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.