missing translation for 'onlineSavingsMsg'
Learn More

6-(2,2,2-Trifluoroethoxy)nicotinic acid, ≥95%, Thermo Scientific™

Product Code. 10243422
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10243422 1 g Amber Glass Bottle 1g
10772123 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10243422 Supplier Thermo Scientific Maybridge Supplier No. RF04860DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175204-90-7
Molecular Formula C8H6F3NO3
Molecular Weight (g/mol) 221.135
MDL Number MFCD00110708
InChI Key GZOOLXWQKURRPU-UHFFFAOYSA-N
Synonym 6-2,2,2-trifluoroethoxy nicotinic acid, 6-2,2,2-trifluoroethoxy pyridine-3-carboxylic acid, 2-trifluoroethoxy pyridine-5-carboxylic acid, 3-pyridinecarboxylic acid, 6-2,2,2-trifluoroethoxy, 6-2,2,2-trifluoro-ethoxy-nicotinic acid, 3-pyridinecarboxylicacid, 6-2,2,2-trifluoroethoxy, maybridge3_005082, pubchem17425, 6-2,2,2 trifluroethoxy nicotinic acid, 6-2,2,2 trifluoroethoxy nicotinic acid
PubChem CID 2781028
IUPAC Name 6-(2,2,2-trifluoroethoxy)pyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)OCC(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.13
Percent Purity 95%
Chemical Name or Material 6-(2, 2, 2-Trifluoroethoxy)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.