missing translation for 'onlineSavingsMsg'
Learn More

6,11-Dihydro-5H-dibenzo[b,e]azepin-6-yl)methanamine Fumarate, Mikromol

Product Code. 30462151
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30462151 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 30462151

Brand: Mikromol MM3383.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6,11-Dihydro-5H-dibenzo[b,e]azepin-6-yl)methanamine Fumarate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6,11-Dihydro-(5H-dibenzo[b,e]azepin-6-yl)methanamine Fumarate
CAS 127785-96-0
Analyte or Component Names 6,11-Dihydro-(5H-dibenzo[b,e]azepin-6-yl)methanamine Fumarate
Shipping Condition Room Temperature
Molecular Formula C15 H16 N2 . C4 H4 O4
Certifications/Compliance ISO 9001
Synonym 5H-Dibenz[b,e]azepine-6-methanamine, 6,11-dihydro-, (2E)-2-butenedioate (1:1): 5H-Dibenz[b,e]azepine-6-methanamine, 6,11-dihydro-, (E)-2-butenedioate (1:1), 6,11-Dihydro-5H-dibenzo[b,e]azepin-6-yl)methanamine fumarate
InChI Formula InChI=1S/C15H16N2.C4H4O4/c16-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)17-15;5-3(6)1-2-4(7)8/h1-8,15,17H,9-10,16H2;1-2H,(H,5,6)(H,7,8)/b;2-1+
Solution Type Neat
SMILES NCC1Nc2ccccc2Cc3ccccc13.OC(=O)\C=C\C(=O)O
IUPAC Name (E)-but-2-enedioic acid;6,11-dihydro-5H-benzo[c][1]benzazepin-6-ylmethanamine
Molecular Weight (g/mol) 340.3731
Quantity 25 mg
Formula Weight 340.1423 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.