missing translation for 'onlineSavingsMsg'
Learn More

5Z-7-Oxozeaenol

Product Code. 15418099
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418099 5 mg 5mg
15428099 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418099 Supplier Thermo Scientific Alfa Aesar Supplier No. J66651.#0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66018-38-0 | C19H22O7 | 362.378 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66018-38-0
Molecular Formula C19H22O7
Molecular Weight (g/mol) 362.378
MDL Number MFCD12912700
InChI Key NEQZWEXWOFPKOT-UHFFFAOYSA-N
Synonym 8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3h-2-benzoxacyclotetradecine-1,7-dione
PubChem CID 53442201
IUPAC Name 5,6,15-trihydroxy-17-methoxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),2,8,15,17-pentaene-7,13-dione
SMILES CC1CC=CC(=O)C(C(CC=CC2=CC(=CC(=C2C(=O)O1)O)OC)O)O

Specifications

Color White
Quantity 5 mg
UN Number UN2811
Solubility Information Soluble in DMSO
Formula Weight 362.37
Physical Form Solid
Chemical Name or Material 5Z-7-Oxozeaenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.