missing translation for 'onlineSavingsMsg'
Learn More

5Z-7-Oxozeaenol, MedChemExpress

Product Code. 16458007
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458007 5 mg 5mg
16451279 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458007 Supplier Medchem Express Supplier No. HY12686
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5Z-7-Oxozeaenol is a natural anti-protozoan compound from fungal origin, acting as a potent irreversible and selective inhibitor of TAK1 and VEGF-R2, with IC50s of 8 nM and 52 nM, respectively.

5Z-7-Oxozeaenol is a natural anti-protozoan compound from fungal origin, acting as a potent irreversible and selective inhibitor of TAK1 and VEGF-R2, with IC50s of 8 nM and 52 nM, respectively.

Specifications

Chemical Name or Material 5Z-7-Oxozeaenol
CAS 253863-19-3
Color White
Purity Grade Notes Research
Molecular Formula C19H22O7
Quantity 5 mg
Synonym FR148083 L783279 LL-Z 1640-2
Solubility Information DMSO : 50 mg/mL (137.98 mM; Need ultrasonic)
SMILES O=C(O[C@@H](C)C/C=C\1)C2=C(O)C=C(OC)C=C2/C=C/C[C@H](O)[C@H](O)C1=O
Molecular Weight (g/mol) 362.37
Formula Weight 362.37
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.