missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5Z-7-Oxozeaenol, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
5Z-7-Oxozeaenol is a natural anti-protozoan compound from fungal origin, acting as a potent irreversible and selective inhibitor of TAK1 and VEGF-R2, with IC50s of 8 nM and 52 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | 5Z-7-Oxozeaenol |
| CAS | 253863-19-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H22O7 |
| Quantity | 1 mg |
| Synonym | FR148083 L783279 LL-Z 1640-2 |
| Solubility Information | DMSO : 50 mg/mL (137.98 mM; Need ultrasonic) |
| SMILES | O=C(O[C@@H](C)C/C=C\1)C2=C(O)C=C(OC)C=C2/C=C/C[C@H](O)[C@H](O)C1=O |
| Molecular Weight (g/mol) | 362.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction