missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5Z)-5-Dodecenoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (5Z)-5-Dodecenoic acid |
| CAS | 2430-94-6 |
| Purity Grade Notes | GC |
| Molecular Formula | C12 H22 O2 |
| Quantity | 50 mg |
| Synonym | (Z)-5-Dodecenoic acid, (5Z)-5-Dodecenoic acid, (Z)-Dodec-5-enoic acid, cis-5-Dodecenoic acid |
| InChI Formula | InChI=1S/C12H22O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h7-8H,2-6,9-11H2,1H3,(H,13,14)/b8-7- |
| SMILES | CCCCCC\C=C/CCCC(=O)O |
| IUPAC Name | (Z)-dodec-5-enoic acid |
| Molecular Weight (g/mol) | 198.3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?