missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse CNTF Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1mg
5 μg
500 μg
Unit Size:
1mg
500µg
5µg
Description
CNTF Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 30 ng/mL, corresponding to a specific activity of >3.3 x 10^4 IU/mg.
Specifications
Specifications
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl, 5 % Trehalose, 0.02% Tween-20. |
| Gene ID (Entrez) | 1270 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 22.5 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of CNTF as determined by LAL method. |
| Gene Alias | ciliary neurotrophic factor, HCNTF |
| Gene Symbol | CNTF |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction