missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 5T4 Monoclonal Antibody (524744)

Description
In direct ELISAs, no cross-reactivity with recombinant mouse 5T4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. The encoded protein is an oncofetal antigen that is specific to trophoblast cells. In adults this protein is highly expressed in many tumor cells and is associated with poor clinical outcome in numerous cancers. Alternate splicing in the 5' UTR results in multiple transcript variants that encode the same protein. [provided by RefSeq, Oct 2009].
Specifications
Specifications
| Antigen | 5T4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 524744 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | TPBG |
| Gene Accession No. | Q13641 |
| Gene Alias | 5T4; 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; 5T4AG; 5T4-AG; AW495680; M6P1; TPBG; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?