missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5S rRNA modificator, MedChemExpress
Description
5S rRNA modificator is a suitable electrophile for 2’-hydroxyl acylation on structured RNA molecules, yielding accurate structural information comparable to that obtained with existing probes
Specifications
Specifications
| Chemical Name or Material | 5S rRNA modificator |
| CAS | 1415238-77-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C9H8N2O2 |
| Quantity | 100 mg |
| Synonym | FAI |
| Solubility Information | DMSO : ≥ 52 mg/mL (295.17 mM) |
| SMILES | O=C(N1C=CN=C1)C2=C(C)OC=C2 |
| Molecular Weight (g/mol) | 176.17 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?