missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (5S,6R)-(+)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone,99%, ee 98%

Product Code. 15415806
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415806 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415806 Supplier Alfa Aesar™ Supplier No. H60498.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 105228-46-4
Melting Point 204°C to 207°C
Color White
Molecular Formula C24H21NO4
MDL Number MFCD00074956
Quantity 1 g
Synonym 2s,3r-benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate, benzyl 2s,3r-+-6-oxo-2,3-diphenyl-4-morpholinecarboxylate, benzyl 2s,3r-6-oxo-2,3-diphenylmorpholine-4-carboxylate, 2s,3r-n-cbz-6-oxo-2,3-diphenylmorpholine, pubchem18127, 2s,3r-+-n-z-6-oxo-2,3-diphenylmorpholine, 2s,3r-benzyl-6-oxo-2,3-diphenylmorpholine-4-carboxylate
InChI Key HECRUWTZAMPQOS-UHFFFAOYNA-N
SMILES O=C(OCC1=CC=CC=C1)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Molecular Weight (g/mol) 387.44
PubChem CID 981233
Formula Weight 387.43
Percent Purity 99%
Physical Form Solid
Chemical Name or Material (5S,6R)-(+)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.