missing translation for 'onlineSavingsMsg'
Learn More

(5RS)-5-Ethyl-2-imino-5-phenyldihydropyrimidine-4,6(1H,3H)-dione (2-Amino-5-ethyl-5-phenyl-4,6(1H,5H)-pyrimidinedione), Mikromol™

Product Code. 30050318
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050318 25 mg 25mg
30049720 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050318 Supplier Mikromol Supplier No. MM0265.130025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(5RS)-5-Ethyl-2-imino-5-phenyldihydropyrimidine-4,6(1H,3H)-dione (2-Amino-5-ethyl-5-phenyl-4,6(1H,5H)-pyrimidinedione)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-5-ethyl-5-phenyl-4,6(1H,5H)-pyrimidinedione
CAS 130690-55-0
Analyte or Component Names 2-Amino-5-ethyl-5-phenyl-4,6(1H,5H)-pyrimidinedione
Shipping Condition Room Temperature
Molecular Formula C12 H13 N3 O2
Certifications/Compliance ISO 17025
Synonym 2-Amino-5-ethyl-5-phenyl-4,6(1H,5H)-pyrimidinedione
InChI Formula InChI=1S/C12H13N3O2/c1-2-12(8-6-4-3-5-7-8)9(16)14-11(13)15-10(12)17/h3-7H,2H2,1H3,(H3,13,14,15,16,17)
SMILES CCC1(C(=O)NC(=N)NC1=O)c2ccccc2
IUPAC Name 5-ethyl-2-imino-5-phenyl-1,3-diazinane-4,6-dione
Molecular Weight (g/mol) 231.25
Quantity 25 mg
Formula Weight 231.1008 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.