missing translation for 'onlineSavingsMsg'
Learn More

(5R,6R)-2,2-Dimethyl-1,3-dioxepane-5,6-diol, Thermo Scientific™

Product Code. 10402294
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Amber Glass Bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10402294 500 mg Amber Glass Bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10402294 Supplier Thermo Scientific Maybridge Supplier No. MAY00284CE
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 61229-03-6
CAS Min % 97.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C7H14O
MDL Number MFCD12198132
Quantity 500 mg
Synonym s-2-pentyloxirane, 2s-2-pentyloxirane, s-1,2-epoxyheptane, 2beta-pentyloxirane, 2s-1,2-epoxyheptane, oxirane,2-pentyl-, 2s, oxirane, 2-pentyl-, 2s
InChI Key NMOFYYYCFRVWBK-ZETCQYMHSA-N
SMILES CCCCCC1CO1
IUPAC Name (2S)-2-pentyloxirane
Molecular Weight (g/mol) 114.188
PubChem CID 10986189
Formula Weight 114.19
Percent Purity 97%
Chemical Name or Material (S)-2-pentyloxirane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.