missing translation for 'onlineSavingsMsg'
Learn More

(5R)-5-Hydroxy-3-oxo-6-(benzyloxy)-hexanoic Acid tert-Butyl Ester, TRC

Product Code. 30396552
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396552 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396552 Supplier Toronto Research Chemicals Supplier No. TRCH953610250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5R)-5-Hydroxy-3-oxo-6-(benzyloxy)-hexanoic Acid tert-Butyl Ester
CAS 147849-63-6
Molecular Formula C17H25O5
Quantity 250 mg
Synonym (5R)-5-Hydroxy-3-oxo-6-(phenylmethoxy)-hexanoic Acid 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C17H24O5/c1-17(2,3)22-16(20)10-14(18)9-15(19)12-21-11-13-7-5-4-6-8-13/h4-8,15,19H,9-12H2,1-3H3/t15-/m1/s1
SMILES CC(C)(C)OC(=O)CC(=O)C[C@@H](O)COCc1ccccc1
IUPAC Name tert-butyl (5R)-5-hydroxy-3-oxo-6-phenylmethoxyhexanoate
Molecular Weight (g/mol) 308.37
Formula Weight 308.1624
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.