missing translation for 'onlineSavingsMsg'
Learn More

(5R)-5-[(1R)-1,2-Dihydroxyethyl]-3,4-dihydroxyfuran-2(5H)-one (D-Isoascorbic Acid), Mikromol™

Product Code. 17957164
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17957164 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17957164 Supplier Mikromol Supplier No. MM0166.100100
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(5R)-5-[(1R)-1,2-Dihydroxyethyl]-3,4-dihydroxyfuran-2(5H)-one (D-Isoascorbic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoascorbic Acid
CAS 89-65-6
Analyte or Component Names Isoascorbic Acid
Shipping Condition Room Temperature
Molecular Formula C6 H8 O6
Certifications/Compliance ISO 9001
Synonym (5R)-5-[(1R)-1,2-Dihydroxyethyl]-3,4-dihydroxyfuran-2(5H)-one (D-Isoascorbic Acid), Ascorbic Acid Impurity F, Sodium Ascorbate Impurity F
InChI Formula InChI=1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5-/m1/s1
SMILES OC[C@@H](O)[C@H]1OC(=O)C(=C1O)O
IUPAC Name (2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Molecular Weight (g/mol) 176.12
Quantity 100 mg
Formula Weight 176.0321 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.