missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylitol-d7, TRC
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Specifications
Specifications
| Chemical Name or Material | Xylitol-d7 |
| Molecular Formula | C5 D7 H5 O5 |
| Quantity | 5 mg |
| Synonym | Xylitol-1,1,2,3,4,5,5-d7, Xylitol-d7, Xylitol-1,1,2,3,4,5,5-D7 |
| InChI Formula | InChI=1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4+,5+/i1D2,2D2,3D,4D,5D |
| SMILES | [2H]C([2H])(O)[C@]([2H])(O)[C@@]([2H])(O)[C@]([2H])(O)C([2H])([2H])O |
| IUPAC Name | (2R,4S)-1,1,2,3,4,5,5-heptadeuteriopentane-1,2,3,4,5-pentol |
| Molecular Weight (g/mol) | 159.189 |
| Formula Weight | 159.112 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction