missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wiskostatin, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Wiskostatin |
| CAS | 253449-04-6 |
| Molecular Formula | C17H18Br2N2O |
| Quantity | 5 mg |
| Synonym | 1-(3,6-Dibromo-9H-carbazol-9-yl)-3-(dimethylamino)propan-2-ol |
| InChI Formula | InChI=1S/C17H18Br2N2O/c1-20(2)9-13(22)10-21-16-5-3-11(18)7-14(16)15-8-12(19)4-6-17(15)21/h3-8,13,22H,9-10H2,1-2H3 |
| SMILES | BrC1=CC2=C(N(CC(CN(C)C)O)C3=C2C=C(Br)C=C3)C=C1 |
| Molecular Weight (g/mol) | 426.15 |
| Formula Weight | 423.9786 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction