missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Voriconazole-d3, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Voriconazole methyl-d3 |
| CAS | 1217661-14-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 2H3 H11 F3 N5 O |
| Quantity | 5 mg |
| Synonym | (αR,βS)-α-(2,4-Difluorophenyl)-5-fluoro-β-(methyl-d3)-α-(1H-1,2,4-triazol-1-ylmethyl)-4-pyrimidineethanol, DRG 0301, UK-109496-d3, VRC-d3, Vfend-d3, Voriconazol-d3 |
| InChI Formula | InChI=1S/C16H14F3N5O/c1-10(15-14(19)5-20-7-22-15)16(25,6-24-9-21-8-23-24)12-3-2-11(17)4-13(12)18/h2-5,7-10,25H,6H2,1H3/t10-,16+/m0/s1/i1D3 |
| SMILES | [2H]C([2H])([2H])[C@@H](c1ncncc1F)[C@](O)(Cn2cncn2)c3ccc(F)cc3F |
| IUPAC Name | (2R,3S)-4,4,4-trideuterio-2-(2,4-difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1,2,4-triazol-1-yl)butan-2-ol |
| Molecular Weight (g/mol) | 352.33 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction