missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin K1 Hydroperoxide, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Vitamin K1 Hydroperoxide |
| CAS | 15576-39-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C31H46O4 |
| Quantity | 5 mg |
| Synonym | 2-(3-Hydroperoxy-3,7,11,15-tetramethyl-1-hexadecen-1-yl)-3-methyl-1,4-naphthalenedione, 2-(3-Hydroperoxy-3,7,11,15-tetramethyl-1-hexadecenyl)-3-methyl-1,4-Naphthoquinone |
| InChI Formula | InChI=1S/C31H46O4/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-20-31(6,35-34)21-19-26-25(5)29(32)27-17-7-8-18-28(27)30(26)33/h7-8,17-19,21-24,34H,9-16,20H2,1-6H3/b21-19+/t23-,24-,31?/m1/s1 |
| SMILES | O=C(C(/C=C/C(C)(OO)CCC[C@H](C)CCC[C@H](C)CCCC(C)C)=C1C)C2=CC=CC=C2C1=O |
| IUPAC Name | 2-[(E,7R,11R)-3-hydroperoxy-3,7,11,15-tetramethylhexadec-1-enyl]-3-methylnaphthalene-1,4-dione |
| Molecular Weight (g/mol) | 482.69 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction