missing translation for 'onlineSavingsMsg'
Learn More

Vitamin B2 (Riboflavin) 13C4 ,15N2, TRC

Product Code. 30457266
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30457266

Brand: Toronto Research Chemicals TRCV6760235MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Riboflavin-2,4,4a,10a-13C4-1,3-15N2
CAS 1217461-14-7
Purity Grade Notes HPLC
Molecular Formula 13C4 C13 H20 15N2 N2 O6
Quantity 5 mg
Synonym Riboflavin-2,4,4a,10a-13C4-1,3-15N2, Riboflavin-13C4,15N2, Vitamin B2-13C4,15N2, 1-Deoxy-1-(3,4-dihydro-7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10(2H)-yl-2,4,4a,10a-13C4-1,3-15N2)-D-ribitol
InChI Formula InChI=1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1/i13+1,15+1,16+1,17+1,19+1,20+1
SMILES Cc1cc2N=[13C]3[13C](=O)[15NH][13C](=O)[15N]=[13C]3N(C[C@H](O)[C@H](O)[C@H](O)CO)c2cc1C
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione
Molecular Weight (g/mol) 382.32
Formula Weight 382.1458
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.