missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vildagliptin Carboxylic Acid Lithium Salt Hydrate, TRC
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
Specifications
Specifications
| Chemical Name or Material | Vildagliptin Carboxylic Acid Lithium Salt Hydrate |
| Analyte or Component Names | Vildagliptin Carboxylic Acid Lithium Salt Hydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17H25LiN2O4 |
| Synonym | N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycyl- L-proline Lithium Salt Hydrate |
| InChI Formula | InChI=1S/C17H26N2O4.Li/c20-14(19-3-1-2-13(19)15(21)22)9-18-16-5-11-4-12(6-16)8-17(23,7-11)10-16;/h11-13,18,23H,1-10H2,(H,21,22);/q;+1/p-1/t11-,12+,13-,16?,17?;/m0./s1 |
| Solution Type | Neat |
| SMILES | O=C(N1[C@H](C(O[Li])=O)CCC1)CN[C@]23C[C@](C[C@H]4C3)(O)C[C@@H](C4)C2 |
| Industry Type | Forensics |
| Molecular Weight (g/mol) | 328.33 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction