missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verapamil-d6, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Verapamil-d6 |
| CAS | 1185193-17-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C27 D6 H32 N2 O4 |
| Quantity | 5 mg |
| Synonym | 2-(3,4-Dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-(1,1,1,3,3,3-hexadeuteriopropan-2-yl)pentanenitrile, Benzeneacetonitrile','α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]methylamino]propyl]-3,4-dimethoxy-α-[1-(methyl-d3)ethyl-2,2,2-d3]-, (±)-Verapamil-d6, Verapamil-D6 (isopropyl-1,1,1,3,3,3-d6), 5-[(3,4-Dimethoxyphenethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-(isopropyl-1,1,1,3,3,3-d6)-valeronitrile, α-[3-[[2-(3,4-Dimethoxyphenyl)ethyl]methylamino]propyl]-3,4-dimethoxy-α-[1-(methyl-d3)ethyl-2,2,2-d3]-benzeneacetonitrile |
| InChI Formula | InChI=1S/C27H38N2O4/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6/h9-12,17-18,20H,8,13-16H2,1-7H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])C(C([2H])([2H])[2H])C(CCCN(C)CCc1ccc(OC)c(OC)c1)(C#N)c2ccc(OC)c(OC)c2 |
| IUPAC Name | 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-(1,1,1,3,3,3-hexadeuteriopropan-2-yl)pentanenitrile |
| Molecular Weight (g/mol) | 460.6386 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction