missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uracil-d4, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Uracil (D4) |
| CAS | 24897-55-0 |
| Molecular Formula | C42H4N2O2 |
| Quantity | 5 mg |
| Synonym | 2,4(1H,3H)-Pyrimidinedione-1,3,5,6-d4 (9CI','ACI), Uracil-1,3,5,6-d4 (8CI), [1,3,5,6-d4]Uracil, Perdeuterouracil, Uracil-d4, Uracil-D4 |
| InChI Formula | InChI=1S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)/i1D,2D/hD2 |
| SMILES | [2H]N1C(=C([2H])C(=O)N([2H])C1=O)[2H] |
| IUPAC Name | 1,3,5,6-tetradeuteriopyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 116.11 |
| Formula Weight | 116.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction