missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenyltin Hydroxide-d15, TRC
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
Specifications
Specifications
| Chemical Name or Material | Triphenyltin Hydroxide-d15 |
| Molecular Formula | C18 D15 H O Sn |
| Quantity | 5 mg |
| Synonym | Hydroxytri(phenyl-d5)stannane, Triphenyltin-d15 hydroxide, Fentin-d15 hydroxide, Fentin hydroxide D15, Hydroxytriphenyltin-d15, Tri(phenyl-d5)(hydroxo)stannane, Triphenylstannyl-d15 hydroxide |
| InChI Formula | InChI=1S/3C6H5.H2O.Sn/c3*1-2-4-6-5-3-1;;/h3*1-5H;1H2;/q;;;;+1/p-1/i3*1D,2D,3D,4D,5D;; |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])[Sn](O)(c2c([2H])c([2H])c([2H])c([2H])c2[2H])c3c([2H])c([2H])c([2H])c([2H])c3[2H] |
| IUPAC Name | hydroxy-tris(2,3,4,5,6-pentadeuteriophenyl)stannane |
| Molecular Weight (g/mol) | 382.122 |
| Formula Weight | 383.116 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction