missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trenbolone-d3, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Trenbolone-d3 |
| CAS | 2026642-62-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18H19D3O2 |
| Quantity | 5 mg |
| Synonym | (17beta)-17-Hydroxyestra-4,9,11-trien-3-one-d3, (+)-Trenbolone-d3, 17beta-Hydroxyestra-4,9,11-trien-3-one-d3, 17beta-Trenbolone-d3, 9,10,11,12-Dehydro-19-nortestosterone-d3, RU 2341-d3, Trenbolone-d3, Trienbolone-d3, beta-Trenbolone-d3 |
| InChI Formula | InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h8-10,15-17,20H,2-7H2,1H3/t15-,16+,17+,18+/m1/s1/i7D2,17D |
| SMILES | [2H]C1([2H])C[C@H]2[C@@H]3CCC4=CC(=O)CCC4=C3C=C[C@]2(C)[C@@]1([2H])O |
| IUPAC Name | (8S,13S,14S,17S)-16,16,17-trideuterio-17-hydroxy-13-methyl-2,6,7,8,14,15-hexahydro-1H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 273.38 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction