missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tranexamic-d2 Acid, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Tranexamic Acid D2 (aminomethyl-D2) |
| CAS | 2714435-89-7 |
| Molecular Formula | C8H13D2NO2 |
| Quantity | 5 mg |
| Synonym | trans-4-(Aminomethyl)cyclohexanecarboxylic-d2 Acid, AMCHA-d2, Amchafibrin-d2, Amikapron-d2, Anvitoff-d2, Bay 3517-d2, CL 65336-d2, Cyclocapron-d2, Cyclokapron-d2, Cyklokapron-d2, DV 79-d2, Exacyl-d2, HAKU-d2, Haematrix-d2, TAMCHA-d2, Tranol-d2, Transamin-d2, Transamin S-d2, Transcam-d2, Ugurol-d2, Xamig-d2, Xed-d2, Zataranax-d2, t-AMCHA-d2, trans-Amcha-d2, trans-p-(Aminomethyl)cyclohexanecarboxylic-d2 Acid |
| InChI Formula | InChI=1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)/t6-,7-/i5D2 |
| SMILES | NC([2H])([2H])[C@@H]1CC[C@@H](C(O)=O)CC1 |
| IUPAC Name | (1r,4r)-4-(aminomethyl-d2)cyclohexane-1-carboxylic acid |
| Molecular Weight (g/mol) | 159.22 |
| Formula Weight | 159.12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction