missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiamethoxam-d3, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Thiamethoxam-d3 |
| CAS | 1294048-82-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 H7 D3 Cl N5 O3 S |
| Quantity | 5 mg |
| Synonym | Thiamethoxam D3 (Methyl D3) |
| InChI Formula | InChI=1S/C8H10ClN5O3S/c1-12-4-17-5-13(8(12)11-14(15)16)3-6-2-10-7(9)18-6/h2H,3-5H2,1H3/i1D3 |
| SMILES | [2H]C([2H])([2H])N1COCN(Cc2cnc(Cl)s2)C1=N[N+](=O)[O-] |
| IUPAC Name | N-[3-[(2-chloro-1,3-thiazol-5-yl)methyl]-5-(trideuteriomethyl)-1,3,5-oxadiazinan-4-ylidene]nitramide |
| Molecular Weight (g/mol) | 294.7332 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction