missing translation for 'onlineSavingsMsg'
Learn More

rac Nebivolol Hydrochloride, TRC

Product Code. 17900755
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17900755 5 mg 5mg
30137595 10 mg 10mg
30132616 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17900755 Supplier Toronto Research Chemicals Supplier No. TRCN3879005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169293-50-9
Molecular Formula C22 H25 F2 N O4 . Cl H
Molecular Weight (g/mol) 441.9
Synonym 2H-1-Benzopyran-2-methanol, α,α'-[iminobis(methylene)]bis[6-fluoro-3,4-dihydro-, hydrochloride (1:1), 2H-1-Benzopyran-2-methanol, α,α'-[iminobis(methylene)]bis[6-fluoro-3,4-dihydro-, hydrochloride (9CI)
IUPAC Name 1-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)-2-[[2-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)-2-hydroxyethyl]amino]ethanol;hydrochloride
SMILES Cl.OC(CNCC(O)C1CCc2cc(F)ccc2O1)C3CCc4cc(F)ccc4O3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H25F2NO4.ClH/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22;/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2;1H
Formula Weight 441.1518
Chemical Name or Material rac Nebivolol, Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.