missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine Sulfoxide, TRC

Product Code. 30003710
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003710 5 mg 5mg
30004789 25 mg 25mg
30003546 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003710 Supplier Toronto Research Chemicals Supplier No. TRCQ5100305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329216-63-9
Molecular Formula C21 H25 N3 O3 S
Molecular Weight (g/mol) 399.51
Synonym 2-[2-[4-(5-Oxidodibenzo[b,f][1,4]thiazepin-11-yl)-1-piperazinyl]ethoxy]ethanol, Quetiapine S-Oxide, Quetiapine Sulfoxide, Quetiapine Fumarate Imp. S (EP)
IUPAC Name 2-[2-[4-(11-oxobenzo[b][1,4]benzothiazepin-6-yl)piperazin-1-yl]ethoxy]ethanol
SMILES OCCOCCN1CCN(CC1)C2=Nc3ccccc3S(=O)c4ccccc24

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H25N3O3S/c25-14-16-27-15-13-23-9-11-24(12-10-23)21-17-5-1-3-7-19(17)28(26)20-8-4-2-6-18(20)22-21/h1-8,25H,9-16H2
Formula Weight 399.1617
Percent Purity >95%
Chemical Name or Material Quetiapine Sulfoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.