missing translation for 'onlineSavingsMsg'
Learn More

MRTX-1257, MedChemExpress

Product Code. 16417587
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16417587

Brand: Medchem Express HY114436

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MRTX-1257 is a selective, irreversible, covalent and orally active KRAS G12C inhibitor, with an IC50 of 900 pM for KRAS dependent ERK phosphorylation in H358 cells.

MRTX-1257 is a selective, irreversible, covalent and orally active KRAS G12C inhibitor, with an IC50 of 900 pM for KRAS dependent ERK phosphorylation in H358 cells.

Specifications

Chemical Name or Material MRTX-1257
Physical Form Solid
CAS 2206736-04-9
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C33H39N7O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 55 mg/mL (97.22 mM; Need ultrasonic)
SMILES N#CC[C@@H]1N(C(C=C)=O)CCN(C2=C3C(CN(C4=C5C(C)=CC=CC5=CC=C4)CC3)=NC(OC[C@H]6N(C)CCC6)=N2)C1
Molecular Weight (g/mol) 565.71
Formula Weight 565.71
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.