missing translation for 'onlineSavingsMsg'
Learn More

MK 677, TRC

Product Code. 30008884
Change view
Click to view available options
Quantity:
5 mg
25 mg
250 mg
Unit Size:
250mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008884 5 mg 5mg
30134073 250 mg 250mg
30011931 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008884 Supplier Toronto Research Chemicals Supplier No. TRCM4249905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159752-10-0
Synonym L-163,191∣MK-0677

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 624.8
Product Type Impurity
Quantity 5 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.