missing translation for 'onlineSavingsMsg'
Learn More

LLY-507, MedChemExpress

Product Code. 16400188
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400188

Brand: Medchem Express HY19313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LLY-507 is a potent and selective inhibitor of protein-lysine methyltransferase SMYD2. LLY-507 potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM. LLY-507 serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes.

LLY-507 is a potent and selective inhibitor of protein-lysine methyltransferase SMYD2. LLY-507 potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM. LLY-507 serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes.

Specifications

Chemical Name or Material LLY-507
CAS 1793053-37-8
Color White
Purity Grade Notes Research
Molecular Formula C36H42N6O
Quantity 5 mg
Solubility Information DMSO : 15 mg/mL (26.10 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC(C2=CC=CC=C2N3CCN(CCN4C=C(C)C5=C4C=CC=C5)CC3)=CC(C#N)=C1)NCCCN6CCCC6
Molecular Weight (g/mol) 574.76
Formula Weight 574.76
Percent Purity 98.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.