missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine-2-d1, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | L-Valine-2-d1 |
| CAS | 77257-03-5 |
| Molecular Formula | C5 D H10 N O2 |
| Quantity | 5 mg |
| Synonym | L-Valine-2-d (9CI','ACI), L-(α-2H)Valine, (2S)-2-Amino-3-methylbutanoic-2-d acid, L-Valine-2-D |
| InChI Formula | InChI=1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1/i4D |
| SMILES | [2H][C@](N)(C(C)C)C(=O)O |
| IUPAC Name | (2S)-2-amino-2-deuterio-3-methylbutanoic acid |
| Molecular Weight (g/mol) | 118.153 |
| Formula Weight | 118.085 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction