missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-[13C5]Xylose, TRC
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
Specifications
Specifications
| Chemical Name or Material | L-[13C5]Xylose |
| Molecular Formula | 13C5H10O5 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/i1+1,2+1,3+1,4+1,5+1 |
| SMILES | O[13C]1[13C@@](O)[13C@](O)[13C@@](O)[13C]O1 |
| Molecular Weight (g/mol) | 155.1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction