missing translation for 'onlineSavingsMsg'
Learn More

(+)-Kavain, MedChemExpress

Product Code. 16451128
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16451128

Brand: Medchem Express HYB16715mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (+)-Kavain, a main kavalactone extracted from Piper methysticum, has anticonvulsive properties, attenuating vascular smooth muscle contraction through interactions with voltage-dependent Na+ and Ca2+ channels. (+)-Kavain is shown to bind at the α4β2δ GABAA receptor and potentiate GABA efficacy. (+)-Kavain is used as a treatment for inflammatory diseases, its anti-inflammatory action has been widely studied.

(+)-Kavain, a main kavalactone extracted from Piper methysticum, has anticonvulsive properties, attenuating vascular smooth muscle contraction through interactions with voltage-dependent Na+ and Ca2+ channels. (+)-Kavain is shown to bind at the α4β2δ GABAA receptor and potentiate GABA efficacy. (+)-Kavain is used as a treatment for inflammatory diseases, its anti-inflammatory action has been widely studied.

Specifications

Chemical Name or Material (+)-Kavain
CAS 500-64-1
Color White
Purity Grade Notes Research
Molecular Formula C14H14O3
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (217.15 mM; Need ultrasonic)
SMILES O=C1C=C(OC)C[C@H](/C=C/C2=CC=CC=C2)O1
Molecular Weight (g/mol) 230.26
Formula Weight 230.26
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.