missing translation for 'onlineSavingsMsg'
Learn More

Imisopasem manganese, MedChemExpress

Product Code. 16408357
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408357 5 mg 5mg
16460196 2 mg 2mg
16453356 25 mg 25mg
16445658 10 mg 10mg
16421827 50 mg 50mg
16416176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16408357 Supplier Medchem Express Supplier No. HY13336

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Imisopasem manganese (M40403) is a stable non-peptidyl mimetic of manganese superoxide MnSOD.

Imisopasem manganese (M40403) is a stable non-peptidyl mimetic of manganese superoxide MnSOD.

Specifications

Chemical Name or Material Imisopasem manganese
Physical Form Solid
CAS 218791-21-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C21H31Cl2MnN5
For Use With (Application) Cancer-programmed cell death
Synonym M40403 GC4403
Solubility Information H2O : 16 mg/mL (33.38 mM; Need ultrasonic and warming)
SMILES [Cl-][Mn+2]1234([Cl-])[N]5=C6C=CC=C5CN1[C@]7([H])CCCC[C@@]7([H])N2CCN3[C@@](CCCC8)([H])[C@]8([H])N4C6
Molecular Weight (g/mol) 479.35
Formula Weight 479.35
Percent Purity 96.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.